missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3e Monoclonal Antibody (FN-18), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen APS0301
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.50000kg
Description
CD3e Monoclonal antibody specifically detects CD3e in Non-human Primate, Rhesus Monkey samples. It is validated for Flow Cytometry.Spécification
CD3e | |
Monoclonal | |
Unconjugated | |
Cd247 | |
4930549J05Rik; A430104F18Rik; AI504783; antigen CD3D, delta polypeptide (TiT3 complex); antigen CD3E, epsilon polypeptide (TiT3 complex); antigen CD3G, gamma polypeptide; antigen CD3Z, zeta polypeptide; AW552088; Cd247; CD247 antigen; CD247 antigen, zeta subunit; Cd247 molecule; CD3; CD3 antigen delta chain; CD3 antigen delta polypeptide; CD3 antigen gamma chain; CD3 antigen, delta polypeptide; CD3 antigen, delta subunit; CD3 antigen, epsilon polypeptide; CD3 antigen, gamma polypeptide; CD3 antigen, zeta polypeptide; CD3 delta; CD3 epsilon; CD3 epsilon chain; CD3 epsilon subunit; CD3 epsilon subunit precursor; CD3 gamma-chain; CD3 glycoprotein; CD3 glycoprotein precursor; CD3 molecule delta polypeptide; CD3 molecule, delta; CD3 molecule, epsilon; CD3 molecule, epsilon polypeptide; CD3 molecule, gamma; CD3 molecule, gamma polypeptide; CD3 protein; CD3 TCR complex; CD3 type I transmembrane glycoprotein; CD3 type I transmembrane glycoprotein precursor; CD3 zeta chain; Cd3d; CD3D antigen d | |
Mouse | |
Ion-exchange chromatography | |
RUO | |
697814, 699467, 699582, 705270 | |
4° C | |
Lyophilized |
Flow Cytometry | |
FN-18 | |
PBS with 125mM trehalose and no preservative | |
0 | |
Cd247, Cd3d, CD3E, CD3g | |
Rhesus monkey peripheral blood leukocytes. | |
500 μg | |
Primary | |
Non-human Primate, Rhesus Monkey | |
Antibody | |
IgG1 |