missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD357 (AITR/GITR) Chimeric Recombinant Rabbit Monoclonal Antibody (YGITR 860.103.5), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen MA547934
358.40 EUR valable jusqu'au 2025-03-16
Utilisez le code promo "24070" pour bénéficier de cette offre.
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Specificity: This antibody binds to mouse GITR.
Tumor necrosis factor receptor superfamily member 18 (TNFRSF18), also called GITR or AITR is a protein that in humans is encoded by the TNFRSF18 gene. It is mapped to 1p36.33. This gene encodes a member of the TNF-receptor superfamily. The encoded receptor has been shown to have increased expression upon T-cell activation, and it is thought to play a key role in dominant immunological self-tolerance maintained by CD25 (+)CD4 (+) regulatory T cells. Knockout studies in mice also suggest the role of this receptor is in the regulation of CD3-driven T-cell activation and programmed cell death. Three alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
Spécification
CD357 (AITR/GITR) Chimeric | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
O35714 | |
Tnfrsf18 | |
Mouse GITR. | |
200 μg | |
Primary | |
Mouse | |
Liquid |
Flow Cytometry, Immunocytochemistry | |
YGITR 860.103.5 | |
Unconjugated | |
Tnfrsf18 | |
Activation-inducible TNFR family receptor; AITR; CD357; GITR; GITR-D; glucocorticoid-induced TNFR-related protein; PRO364; TNF receptor superfamily activation-inducible protein; TNF receptor superfamily member 18; TNFRSF18; Tumor necrosis factor receptor superfamily member 18; tumor necrosis factor receptor superfamily, member 18; UNQ319; UNQ319/PRO364 | |
Rabbit | |
Affinity chromatography | |
RUO | |
21936 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG κ |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
CD357 (AITR/GITR) Chimeric Recombinant Rabbit Monoclonal Antibody (YGITR 860.103.5), Invitrogen™ > 200 μg; Unconjugated
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu