missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34 Mouse anti-Human, Oligo, Clone: 581, BD™ AbSeq ™
Mouse Monoclonal Antibody
Marque: BD Biosciences 940021
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD34 Monoclonal antibody specifically detects CD34 in Human samples. It is validated for Single Cell 3' SequencingSpécification
CD34 | |
Monoclonal | |
Oligo | |
gp105-120; My10; Hematopoietic progenitor cell antigen CD34 | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
Single Cell 3' Sequencing | |
Clone 581 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
RUO | |
Human | |
IgG1 κ |