missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34 Mouse anti-Human, Clone: 3C8G12, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 60180-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD34 is a highly glycosylated monomeric with a molecular weight range of 111-115 kDa surface protein that is present on many stem cell populations. CD34 is a stem cell marker although its expression on human hematopoietic stem cells is reversible. CD34 may serve as a surface receptor that undergoes receptor-mediated endocytosis and regulates adhesion, differentiation and proliferation of hematopoietic stem cells and other progenitors. CD34 expression is likely to represent a specific state of hematopoietic development that may have altered adhering properties with expanding and differentiating capabilities in both in vitro and in vivo conditions. CD34 is possibly an adhesion molecule with a putative role for mediating the attachment of stem cells to the bone marrow extracellular matrix or directly to stromal cells. Further, CD34 could act as a scaffold for the attachment of lineage specific glycans, allowing stem cells to bind to lectins expressed by stromal cells or other marrow components. CD34 is thought to have a role in presenting carbohydrate ligands to selectins. The intracellular chain of the CD34 antigen is a site of phosphorylation by activated protein kinase C suggesting a putative role in signal transduction. Diseases associated with CD34 dysfunction include dermatofibrosarcoma and neurofibroma.Spécification
| CD34 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P28906 | |
| CD34 | |
| CD34 Fusion Protein Ag5996 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 3C8G12 | |
| Unconjugated | |
| CD34 | |
| CD34, CD34 molecule | |
| Mouse | |
| Caprylic Acid Precipitation | |
| RUO | |
| 947 | |
| -20°C | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu