missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD177 Monoclonal Antibody (MEM-166), APC, Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA110119
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD177 (NB1/HNA-2a and PRV-1 form) is a GPI-anchored glycoprotein present mainly on neutrophils. Its plasma membrane expression is increased during pregnancy and and inflammation or after G-CSF application. Ligand of CD177 has been identified as CD31 (PECAM-1). CD177 participates in neutrophil transmigration and seems to be also a pro-proliferative molecule. The antibodies against CD177 can be involved in neonatal alloimmune neutropenia (NAN).Spécification
CD177 | |
Monoclonal | |
APC | |
CD177 | |
1190003K14Rik; CD177; CD177 antigen; CD177 molecule; cell surface receptor; HNA2A; HNA-2a; Human neutrophil alloantigen 2a; NB1; NB1 glycoprotein; NB1 GP; Pdp3; Polycythemia rubra vera protein 1; PRV1; PRV-1; RGD1562941; UNQ595/PRO1181 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
57126 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-166 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
Q8N6Q3 | |
CD177 | |
Human granulocytes | |
100 Tests | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu