missing translation for 'onlineSavingsMsg'
Learn More

CD16 Mouse anti-Human, PE, Clone: 3G8, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 560995

Code nomenclature Nacres: NA.41

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15860669

  • 193.00€ / 1 pièce
Expédition estimée: 20-09-2024
pour voir le stock disponible 

Description

Description

Reacts with the 50-65kDa transmembrane form of IgG Fc (FcγRIII), a human NK-cell-associated antigen. CD16 is expressed on NK cells as well as macrophages and granulocytes. Reports indicate that CD16 plays a role in signal transduction and NK cell activation. CD16 antibody blocks the binding of soluble immune complexes to granulocytes. This clone also cross-reacts with a subset of peripheral blood lymphocytes and monocytes, but not granulocytes, of baboon and both rhesus and cynomolgus macaque monkeys. Multi-color analysis reveals that the distribution on lymphocytes is similar to that found in human studies, with the majority of CD16-positive lymphocytes being both CD3 and CD20 negative. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: 3G8
Isotype: IgG1, κ
Cross Reactivity: Human, Rhesus, Cynomolgus, Baboon

Flow Cytometry

Spécification

Spécification

CD16
Monoclonal
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human polymorphonuclear leukocytes
25 Tests
Immunology
Baboon, Cynomolgus, Human, Rhesus Monkey
IgG1 κ
Flow Cytometry
3G8
FcRIII; Fc-gamma RIII; FCG3; FCGR3; FCGRIII; FcγRIII; IGFR3
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

For Research Use Only.