Learn More
Carbonic Anhydrase XIV Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marque: Invitrogen PA547270
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant mouse CA14 is observed and less than 1% cross-reactivity with recombinant human CA1, 2, 3, 4, 8, 9, 10, 12, and 13 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Carbonic Anhydrase XIV Polyclonal antibody specifically detects Carbonic Anhydrase XIV in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot.Spécification
Carbonic Anhydrase XIV | |
Polyclonal | |
Unconjugated | |
CA14 | |
AW536446; CA XIV; CA14; Car14; Carbonate dehydratase XIV; carbonic anhydrase 14; Carbonic anhydrase XIV; carbonic dehydratase; Catm; CAXiV; CA-XIV; UNQ690/PRO1335 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
23632, 23831 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9ULX7, Q9WVT6 | |
CA14, Car14 | |
Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase XIV/CA14 Gly19-Met290 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |