missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase VIII Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA542120
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Peptide sequence: TISQLQIEEF RRLRTHVKGA ELVEGCDGIL GDNFRPTQPL SDRVIRAAFQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
Carbonic Anhydrase VIII Polyclonal antibody specifically detects Carbonic Anhydrase VIII in Human samples. It is validated for Western Blot.Spécification
Carbonic Anhydrase VIII | |
Polyclonal | |
Unconjugated | |
Ca8 | |
AW546993; Ca8; Cals; Cals1; CAMRQ3; Car8; carbonate dehydratase; carbonic anhydrase 8; carbonic anhydrase VIII; carbonic anhydrase-like sequence; carbonic anhydrase-like sequence 1; carbonic anhydrase-related protein; CA-related protein; CARP; CA-RP; CARP VIII; CA-RP VIII; CA-VIII; hypothetical protein LOC550233; LOW QUALITY PROTEIN: carbonic anhydrase-related protein; MGC120502; MGC99509; wdl; zgc:110118 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
767 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P35219 | |
Ca8 | |
Synthetic peptide directed towards the middle region of human CA8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |