missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase III Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marque: Invitrogen PA547265
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) CA1, rhCA2, rhCA4, rhCA9, rhCA10, rhCA13 and rhCA14 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Carbonic Anhydrase III Polyclonal antibody specifically detects Carbonic Anhydrase III in Human, Mouse, Rat samples. It is validated for Immunoprecipitation, Western Blot.Spécification
Carbonic Anhydrase III | |
Polyclonal | |
Unconjugated | |
Ca3 | |
BB219044; CA III; Ca3; CAH3; CAIII; CA-III; CAIII (Muscle); Car3; Car-3; Carbonate dehydratase III; carbonic anhydrase 3; Carbonic anhydrase III; carbonic anhydrase III, muscle specific; carbonic anhydrase-like protein; EC 4.2.1.1; epididymis secretory sperm binding protein Li 167mP; HEL-S-167mP | |
Goat | |
Antigen affinity chromatography | |
RUO | |
12350, 54232, 761 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P07451, P14141, P16015 | |
Ca3, Car3 | |
E. coli-derived recombinant human Carbonic Anhydrase III/CA3 Ala2-Lys260 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |