missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPSL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA563579
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: PMSRARKEVI MQAFRKLDKT GDGVITIEDL REVYNAKHHP KYQNGEWSEE Q Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
CAPSL gene ontology annotations related to this gene include calcium ion binding.Spécification
CAPSL | |
Polyclonal | |
Unconjugated | |
Capsl | |
1700028N11Rik; AW125205; Calcyphosine; calcyphosine like; calcyphosine-like; calcyphosin-like protein; CAPSL; RGD1308776 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
133690 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WWF8 | |
Capsl | |
Recombinant protein corresponding to Human CAPSL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only