missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C20orf4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 20179-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
AAR2 encodes the homolog of the yeast A1-alpha2 repressin protein that is involved in mRNA splicing. Alternately spliced transcript variants have been found for this gene.Spécification
C20orf4 | |
Polyclonal | |
Unconjugated | |
AAR2 | |
C20orf4, CGI 23, PRO0225 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25980, 296312, 68295 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9D2V5, Q9Y312 | |
Aar2 | |
C20orf4 Fusion Protein Ag14039 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |