Learn More
Motic™ C-Mount Camera Adapter for SMZ-160
The C-Mount Camera Adapter is compatible with the SMZ-160 microscope.
Marque: Motic™ 1101010100881
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- C-mount camera adapter for SMZ-160.
- Available in 0.35x, 0.5x, or 1.0x (no lens) magnification.
- Adapt 1/3 in. or 1/2 in. chip sensors.
Spécification
Adapter | |
C-Mount | |
To use with a digital camera for microscopy | |
1.0X | |
5 years |
Non autoclavable | |
To screw a digital camera on a trinocular SMZ-160 | |
No Lens | |
Non stérile |