missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromoéthane, 98 %, Thermo Scientific Chemicals
Click to view available options
Quantité:
500 mL
1 L
2.5 L
Conditionnement:
Flacon en verre
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Réactif d’alkylation
Spécification
Spécification
Nom chimique ou matériau | Bromoethane |
Note relative au nom | Pure |
Couleur | Incolore à jaune |
Spectre IR | Authentic |
Plage de pourcentage du dosage | 98% |
Conditionnement | Flacon en verre |
Formule linéaire | C2H5Br |
Indice de réfraction | 1.4230 to 1.4250 (20°C, 589 nm) |
Quantité | 2.5 L |
Beilstein | 01, 88 |
Afficher plus |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'