missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRCA2 Antibody (5F6), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Marque: Novus Biologicals NBP1-41189
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
BRCA2 Monoclonal specifically detects BRCA2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Spécification
BRCA2 | |
Monoclonal | |
1.0mg/mL | |
Western Blot 1-5 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 | |
BRCA1/BRCA2-containing complex, subunit 2, BRCC2, breast and ovarian cancer susceptibility gene, early onset, breast cancer 2 tumor suppressor, breast cancer 2, early onset, breast cancer susceptibility protein BRCA2, breast cancer type 2 susceptibility protein, BROVCA2, FACD, FAD, FAD1, FANCB, FANCD, FANCD1Fanconi anemia, complementation group D1, Fanconi anemia group D1 protein, GLM3, PNCA2 | |
Mouse | |
384 kDa | |
100 μL | |
Breast Cancer, Cancer | |
675 | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
5F6 | |
Unconjugated | |
P51587 | |
BRCA2 | |
Amino acids 2336-2478 of human BRCA2 expressed in E. coli. BRCA2-P400. | |
Immunogen affinity purified | |
RUO | |
Primary | |
This antibody will recognize human BRCA2. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
For Research Use Only