Learn More
Bovine Serum Albumin Antibody (ALB/398), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Marque:  Novus Biologicals NBP2-76991B
Code nomenclature Nacres: NA.46
Code produit. 18604701
  • 629.00€ / 0.10mL
Description
Bovine Serum Albumin Monoclonal antibody specifically detects Bovine Serum Albumin in Bovine, Canine (Negative), Guinea Pig (Negative), Human (Negative), Rabbit (Negative) samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Bovine Serum Albumin
Monoclonal
Biotin
albumin, BSA, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Purified Bovine Serum Albumin (Uniprot: P02769)
0.1 mL
Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers
280717
Store at 4°C in the dark.
IgG1 κ
ELISA
ALB/398
PBS
Mouse
Protein A or G purified
RUO
Primary
Bovine, Canine (Negative), Guinea Pig (Negative), Human (Negative), Rabbit (Negative)
Purified
Suggestions de produits
Suggestions de produits
Vidéos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.