Learn More
Robu™ Entonnoirs de filtration Buchner en verre borosilicaté
Robu™ Büchner Filter Funnels with Glass Filter Discs are made of pure borosilicate 3.3 glass and are highly resistant to most chemicals, including solvents, acids, and alkalis. The VitraPOR™ filter disc is rated as Porosity 5 (Pore size: 1 to 1.6 μm, Standard: ISO 4793-80).
Marque: Robu™ 21505
Code nomenclature Nacres: NB.43
Informations supplémentaires : Poids : 0.10000kg
Description
- Product Type: Büchner Filter Funnel
- Material: Borosilicate 3.3 Glass (Body), VitraPOR™ Glass (Filter)
- Porosity: Grade 5
- Pore Size: 1-1.6 μm
- Standard: ISO 4793-80
- For Use With (Application): Filtration
- For Use With (Equipment): Guko Rubber Gaskets (e.g. 10430652)
Spécification
40 mm (dia. ext.) ; 36 mm (dia. int.) | |
10mm | |
53 mm | |
Très résistant à la plupart des produits chimiques tels que les solvants, les acides et les alcalins | |
Guko Rubber Gaskets (e.g. 10430652) | |
1.7 oz. |
50 ml | |
Verre borosilicaté | |
ISO 4793-80 | |
40mm | |
1 to 1.6 μm | |
Entonnoir de filtration Buchner |