missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Bipolar Pulse Switcher
Easily execute automatic grouped pulse, bipolar switching protocols by adding the Bipolar Pulse Switcher to your ECM 830 or Gemini X2 electroporation waveform generator.
Marque: BTX™ 452061
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Pulse switcher protocols allow a user to create and run protocols containing groups of square wave pulses with different polarity, pulse number, pulse length, and pulse interval characteristics.
- Pulse Switcher User Defined Protocols may be programmed with up to 10 groups of square wave pulses with separately defined polarity (+ or -) for each group.
- The user may thus incorporate bipolar switching into their electroporation protocols.
- The maximum number of pulses per group in a Pulse Switcher User Defined Protocol is 100 pulses.
- The number of pulses, pulse length, and pulse interval is separately defined for each group of pulses in Pulse Switcher User Defined Protocols.
- The minimum interval between pulses and between groups of pulses for Pulse Switcher User Defined Protocols is 50 msec.
- The maximum number of pulses per Pulse Switcher User Defined Protocol is 1,000 pulses.
Applications:
- In Vitro Suspension
- Adherent Cell Transfection
- In Vivo, In Ovo and In Utero Gene and Drug Delivery
- Explant Tissue Transfection
- CRISPR Gene Editing
Spécification
Cell Transfection, Gene and Drug Delivery, Tissue Transfection, CRISPR Gene Editing | |
BTX Electroporation | |
100/240 V | |
CE, WEEE, RoHS | |
Pulse Switcher, Power Supply, and USB Connector Cable | |
5°C à 40°C | |
0.6 lb. | |
Non-sterile | |
505 to 3000 V (in 5 V Steps) | |
Fiche banane avec gaine rétractable | |
Onde carrée |
Gemini X2 and ECM830 Electroporators | |
Système d’électroporation | |
Color Touchscreen | |
Bipolar | |
41°F à 104°F | |
20% to 80% RH, Non-condensing | |
276 g | |
Suitable for All Cell Types | |
5 to 500 V (in 1 V Steps) | |
Digital User |