Learn More
Beta Actin Mouse anti-Human, HRP, Clone: 2D4H5, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech HRP-66009-100UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The Beta Actin antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human Beta Actin. This antibody recognizes human, mouse, rat, hamster, zebrafish, monkey antigen. The Beta Actin antibody has been validated for the following applications: WB analysis. This antibody is hrp.
Spécification
Beta Actin | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Hamster, Zebrafish |
Western Blot | |
2D4H5 | |
HRP | |
100 μL | |
60 | |
IgG2b |