Learn More
Beta Actin Mouse anti-Human, HRP, Clone: 2D4H5, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech HRP-66009-100UL
Code produit. 16891246
  • 189.00€ / 100µL
Découvrez plus d'offres spéciales
Description
Description
The Beta Actin antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human Beta Actin. This antibody recognizes human, mouse, rat, hamster, zebrafish, monkey antigen. The Beta Actin antibody has been validated for the following applications: WB analysis. This antibody is hrp.
Spécification
Spécification
Beta Actin
Monoclonal
500 μg/mL
Mouse
Primary
Human, Mouse, Rat, Hamster, Zebrafish
Western Blot
2D4H5
HRP
100 μL
60
IgG2b