Learn More
Berkshire™ BCR™ Small Aluminum Mop Frame
Mop System replicates the most effective way to clean a surface- wiping. Easy to use, highly effective and quick. Flat Mop Head is light and easy to maneuver, even with wet wipers attached. Clean, light and durable.
Marque: Berkshire™ BCR.MOP3.1
Code nomenclature Nacres: NB.34
Informations supplémentaires : Poids : 0.26000kg
Description
BCR™ Small Aluminum Mop Frame replicates the most effective way to clean a surface- wiping. Easy to use, highly effective and quick. Flat Mop Head is light and easy to maneuver, even with wet wipers attached. Clean, light and durable.
- Clean, light and durable
- High quality, anodized aluminum
- Autoclavable
- Plastic eyes for wiper attachment
Specifications
Flat Mop | |
Autoclavable | |
Anodized Aluminum |
Non-sterile | |
No |