missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAT3/BAG6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP1-18927
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
BAT3/BAG6 Polyclonal specifically detects BAT3/BAG6 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Spécification
| BAT3/BAG6 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| BAT3HLA-B associated transcript-3, BCL2-associated athanogene 6BAG-6, D6S52EHLA-B-associated transcript 3, G3BAG family molecular chaperone regulator 6, HLA-B associated transcript 3, large proline-rich protein BAT3, Protein G3, Protein Scythe, scythe | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 7917 | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot, Immunoprecipitation 1:10-1:500 | |
| NP_004630.3 | |
| BAG6 | |
| The immunogen recognized by this antibody maps to a region between residue 200 and 250 of human HLA-B associated transcript 3 using the numbering given in entry NP_004630.3 (GeneID 7917). | |
| 0.1 mL | |
| Primary | |
| Human. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Horse, Rhesus Monkey, Gorilla, Crab-eating macaque, Chinese hamster and Northern white-cheeked gibbon. | |
| Store at 4C. Do not freeze. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu