missing translation for 'onlineSavingsMsg'
Learn More

BAP1 Antibody (1G8), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication

226.00€ - 517.00€

Spécification

Antigène BAP1
Clone 1G8
Concentration 1 mg/mL
Dilution Western Blot 2 ug/ml, Flow Cytometry 1 ug per million cells
Applications Western Blot, Flow Cytometry
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
18265470
View Documents
Novus Biologicals
NB110-60521SS
0.025 mL
226.00€
25µL
View alternative products
Save Up to 
»
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
18796963
View Documents
Novus Biologicals
NB110-60521
517.00€
0.10mL
View alternative products
Save Up to 
»
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

BAP1 Monoclonal specifically detects BAP1 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

BAP1
1 mg/mL
Western Blot, Flow Cytometry
Unconjugated
Mouse
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Tumor Suppressors
Q92560
8314
Full-length human BAP1 protein expressed and purified from E.coli. [UniProt# Q92560]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1G8
Western Blot 2 ug/ml, Flow Cytometry 1 ug per million cells
Monoclonal
Purified
RUO
Human
BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase), BRCA1-associated protein 1, Cerebral protein 6, cerebral protein-13, cerebral protein-6, DKFZp686N04275, EC 3.4.19.12, FLJ35406, FLJ37180, hucep-6, KIAA0272HUCEP-13, ubiquitin carboxyl-terminal hydrolase BAP1, ubiquitin carboxy-terminal hydrolase, UCHL2
BAP1
IgG1 κ
Protein G purified
85 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.