missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAMB-4, MedChemExpress
Click to view available options
Quantité:
5 mg
10 mg
Description
BAMB-4(ITPKA-IN-C14) is a new membrane-permeable inhibitor against inositol-1,4,5-trisphosphate-3-kinase A((ITPKA) with IC50 of 37 μM in ADP-Glo Assay.
Specifications
Specifications
| Nom chimique ou matériau | BAMB-4 |
| Forme physique | Solid |
| CAS | 891025-25-5 |
| Couleur | Off-White |
| Quantité | 5 mg |
| Notes de qualité de la pureté | Research |
| Formule moléculaire | C15H12N2O2 |
| À utiliser avec (application) | Cancer-Kinase/protease |
| Synonyme | ITPKA-IN-C14 |
| Informations sur la solubilité | DMSO : ≥ 40 mg/mL (158.56 mM) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction