missing translation for 'onlineSavingsMsg'
Learn More

AV-412 free base, MedChemExpress

Code produit. 16434886
Encompass_Preferred
Click to view available options
Quantité:
5 mg
10 mg
50 mg
100 mg
This item is not returnable. View return policy

Product Code. 16434886

missing translation for 'mfr': Medchem Express HY10346A

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

MedChemExpress AV-412 free base (MP-412 free base) is an EGFR inhibitor with IC50s of 0.75, 0.5, 0.79, 2.3, 19 nM for EGFR, EGFRL858R, EGFRT790M, EGFRL858R/T790M and ErbB2, respectively.

AV-412 free base (MP-412 free base) is an EGFR inhibitor with IC50s of 0.75, 0.5, 0.79, 2.3, 19 nM for EGFR, EGFRL858R, EGFRT790M, EGFRL858R/T790M and ErbB2, respectively.

Specifications

Nom chimique ou matériau AV-412 free base
CAS 451492-95-8
Couleur Jaune
Notes de qualité de la pureté Research
Formule moléculaire C27H28ClFN6O
Quantité 100 mg
Synonyme MP-412 free base
Informations sur la solubilité DMSO : 50 mg/mL (98.62 mM; Need ultrasonic)
SMILES O=C(C=C)NC1=CC2=C(NC3=CC=C(C(Cl)=C3)F)N=CN=C2C=C1C#CC(C)(C)N(CC4)CCN4C
Poids moléculaire (g/mol) 507.0
Poids de la formule 507.0
Pourcentage de pureté 98.33%
Qualité Research
Forme physique Solid
À utiliser avec (application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.