missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATPG Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-15525
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
ATPG Polyclonal specifically detects ATPG in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Spécification
| ATPG | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| ATP synthase gamma chain, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1, ATP5CATP synthase subunit gamma, mitochondrial, ATP5CL1mitochondrial ATP synthase, gamma subunit 1, F-ATPase gamma subunit | |
| Rabbit | |
| 33 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Zebrafish | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.44mg/mL | |
| Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| P36542 | |
| ATP5F1C | |
| Recombinant protein encompassing a sequence within the center region of human ATP synthase gamma. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 509 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu