missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASRGL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 11400-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
ASRGL1 gene ontology annotations related to this gene include asparaginase activity.Spécification
ASRGL1 | |
Polyclonal | |
Unconjugated | |
ASRGL1 | |
ALP, ALP1, asparaginase like 1, Asparaginase like protein 1, ASRGL1, CRASH, L asparaginase, L asparagine amidohydrolase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
246307, 66514, 80150 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7L266, Q8C0M9, Q8VI04 | |
Asrgl1 | |
ASRGL1 Fusion Protein Ag1955 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |