missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASCC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17627-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene encodes a protein that belongs to a family of helicases that are involved in the ATP-dependent unwinding of nucleic acid duplexes. The encoded protein is the largest subunit of the activating signal cointegrator 1 complex that is involved in DNA repair and resistance to alkylation damage. Alternate splicing results in multiple transcript variants.Spécification
ASCC3 | |
Polyclonal | |
Unconjugated | |
ASCC3 | |
ASC 1 complex subunit p200, ASC1p200, ASCC3, HELIC1, Helicase, ATP binding 1, RNAH, Trip4 complex subunit p200 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10973, 309887, 77987 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
E9PZJ8, F1LPQ2, Q8N3C0 | |
Ascc3 | |
ASCC3 Fusion Protein Ag11848 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
ASCC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu