Learn More
ARL13B Mouse anti-Human, CoraLite 594, Clone: 1H6C3, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech CL594-66739-100UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The ARL13B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ARL13B. This antibody recognizes human, canine antigen. The ARL13B antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Spécification
ARL13B | |
Monoclonal | |
2000 μg/mL | |
Mouse | |
Primary | |
Human, Canine |
Immunofluorescence | |
1H6C3 | |
CoraLite 594 | |
100 μL | |
200894 | |
IgG2a |