Learn More
APOE Mouse anti-Human, Clone: OTI2A4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marque: Origene Technologies CF805331
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Apo E (Apolipoprotein E) plays an important role in the metabolism of lipids in the plasma, and is also is a constituent of various plasma lipoprotein-lipid particles. The protein is synthesized in liver, brain, spleen, and the kidney, and concentrations are high in interstitial fluids, where it plays a role in distributing cholesterol from cells where cholesterol is in excess to cells which require cholesterol. Apo E also plays an important role in the formation of very low density lipoprotein and chylomicrons. The Apo E gene is located on chromosome 19 at position 19q13.2.Spécification
| APOE | |
| Monoclonal | |
| Unconjugated | |
| APOE | |
| AD2, APO-E, ApoE4, LDLCQ5, LPG | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 348 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P02649 | |
| APOE | |
| Full length human recombit protein of human APOE produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.