missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP2 alpha Antibody (AP2a 8G8/5), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NB100-2639H
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
AP2 alpha Monoclonal specifically detects AP2 alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Spécification
| AP2 alpha | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| TFAP2A | |
| Bacterialy expressed AP-2a/AP-2g fusion protein. | |
| 0.1 mL | |
| Cancer, Transcription Factors and Regulators | |
| 7020 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| AP2a 8G8/5 | |
| Western Blot, Immunohistochemistry | |
| Activating enhancer-binding protein 2-alpha, Activator protein 2, AP-2 transcription factor, AP2-alpha, AP2TFAP-2alpha, AP-2transcription factor AP-2 alpha (activating enhancer-binding protein 2 alpha), BOFS, FLJ51761, TFAP2, transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha), transcription factor AP-2-alpha | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| AP2a 8G8/5 is specific for the alpha form of AP-2.Antigen recovery using microwave or pressure cooking required for paraffin embedded tissue. | |
| Store at 4C in the dark. | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu