Learn More
XBP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA527650
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, HepG2 (3 μM Thapsigargin treatment for 24 hr). Predicted reactivity: Pig (84%), Rhesus Monkey (98%), Bovine (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
XBP1 Polyclonal antibody specifically detects XBP1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spécification
XBP1 | |
Polyclonal | |
Unconjugated | |
XBP1 | |
D11Ertd39e; Hepatocarcinogenesis-related transcription factor; HTF; OTTHUMP00000028682; Tax-responsive element-binding protein 5; tax-responsive element-binding protein 5 homolog; TREB5; TREB-5; X-box binding protein 1; X-box-binding protein 1; X-box-binding protein 1, cytoplasmic form; X-box-binding protein 1, luminal form; XBP1; XBP-1; XBP2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7494 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.93 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P17861 | |
XBP1 | |
Recombinant fragment corresponding to a region within amino acids 90 and 261 of Human XBP1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |