missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WAC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA534536
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
A suggested positive control is human testis tissue lysate. PA5-34536 can be used with blocking peptide PEP-1579.
The WW domain containing adaptor with coiled-coil protein (WAC) contains a WW domain that mediates protein-protein interactions and colocalizes with RNA splicing factor SC35. Further studies have indicated that WAC is a functional partner of the RNF20/40 complex that ubiquitinates Histone H2B, and that WAC regulates H2B ubiquitination. WAC targets RNF20/40 to associate with RNA polymerase II complex for H2B ubiquitination at active transcription sites. WAC-dependent transcription is also important for cell-cycle checkpoint activation in response to genotoxic stress.Spécification
WAC | |
Polyclonal | |
Unconjugated | |
Wac | |
1110067P07Rik; A230035H12Rik; AI256735; AI256776; BM-016; DESSH; Kiaa1844; PRO1741; RGD1562407; Wac; WW domain containing adaptor with coiled-coil; WW domain-containing adapter protein with coiled-coil; WW domain-containing adaptor with coiled-coil; WW domain-containing protein 4; Wwp4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
225131, 307029, 51322 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q924H7, Q9BTA9 | |
Wac | |
A 19 amino acid peptide near the center of human WAC | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu