missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCL1A, Human, IP-WB Antibody Pair (PW1), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Marque: Abnova H00008115-PW1.1Set
Includes: Antibody pair for IP: mouse monoclonal anti-TCL1A (300μL), Antibody pair for WB: rabbit polyclonal anti-TCL1A (50μL)
Spécification
TCL1A | |
Monoclonal/Polyclonal | |
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
TCL1 | |
Mouse, Rabbit | |
RUO | |
8115 | |
Human | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
TCL1A | |
TCL1A | |
1 Set | |
Primary | |
Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-TCL1A (300 ul) 2. Antibody pair for WB: rabbit polyclonal anti-TCL1A (50 ul) | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
For Research Use Only