Learn More
Ran Mouse, Unlabeled, Clone: 20, BD
Mouse Monoclonal Antibody
345.00€ - 520.00€
Spécification
Antigène | Ran |
---|---|
Clone | 20 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15845409
|
BD Biosciences
610340 |
50 μg |
345.00€
1 kit |
Expédition estimée: 20-09-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15855409
|
BD Biosciences
610341 |
150 μg |
520.00€
1 kit |
Expédition estimée: 20-09-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Ran is a highly conserved GTPase that is ubiquitously expressed. At steady-state, 80-90% of cellular Ran is located in the nucleus, with the remainder in the cytoplasm. The human TC4/Ran cDNA was originally isolated based on the predicted homology of its encoded protein product to Ras. Unlike other members of the Ras superfamily, Ran does not appear to contain signals for lipid modification, nor does it appear to show membrane localization. Ran has been implicated in a number of cellular processes such as the initiation of DNA replication, entry into and exit from mitosis and in nuclear RNA and protein transport through the nuclear pore complex. A number of Ran binding proteins have been identified. One of the smaller proteins identified is a 28kDa cytosolic molecule known as RanBP1. The GTP-bound state of Ran is stabilized through its binding to RanBP1.
Spécification
Ran | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG2a | |
Affinity Purified |
20 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Chicken, Human, Mouse, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.