Learn More
NFAT-1 Mouse, Unlabeled, Clone: 1, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 610702
Description
T cells are activated and induced to proliferate following binding of their respective antigen. The process of includes expression of genes that encode factors (i.e., cytokines) which regulate various cell types. Modulation of gene expression is conducted by an array of specific interactions between transcription factors and DNA. NFAT-1 (Nuclear Factor of Activated T cells) is a transcription factor that regulates expression of the interleukin-2 gene. Thus, NFAT-1 DNA binding activity is undetectable in resting cells, but increases during T-cell activation. NFAT-1, a protein of 921 amino acids, is part of an oligomeric transcription factor that also contains Fra-1 and JunB. NFAT-1 was initially described as a phosphoprotein and is dephosphorylated in activated T cells transformed with the leukemia virus HTLV-l.
Spécification
| NFAT-1 | |
| Monoclonal | |
| 250μg/mL | |
| NF-ATc2; NFATP; KIAA0611 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
| Western Blot | |
| 1 | |
| Unconjugated | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| 50 μg | |
| Cell Biology | |
| Canine, Human | |
| IgG1 |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
For Research Use Only.