missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monoacylglycerol Lipase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA527915
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Recommended positive controls: HeLa, HepG2, mouse adipose. Predicted reactivity: Mouse (84%), Rat (84%), Pig (91%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Monoglyceride lipase functions together with hormone-sensitive lipase to hydrolyze intracellular triglyceride stores in adipocytes and other cells to fatty acids and glycerol. MGLL may also complement lipoprotein lipase in completing hydrolysis of monoglycerides resulting from degradation of lipoprotein triglycerides.Spécification
Monoacylglycerol Lipase | |
Polyclonal | |
Unconjugated | |
MGLL | |
AA589436; HUK5; HU-K5; lysophospholipase homolog; Lysophospholipase-like; MAGL; Mgl; Mgl2; MGLL; Monoacylglycerol lipase; monoglyceride lipase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11343, 23945, 29254 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
O35678, Q8R431, Q99685 | |
MGLL | |
Recombinant fragment corresponding to a region within amino acids 36 and 303 of Human Monoglyceride lipase | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |