Learn More
Ly-6G Rat anti-Mouse, Alexa Fluor 700, Clone: 1A8, BD
Rat Monoclonal Antibody
Marque: BD Biosciences 561236
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
The 1A8 monoclonal antibody specifically binds to Ly-6G, a 21-25-kDa GPI-anchored protein. In the bone marrow, Ly6G is expressed on the majority of the largest cells, that are predominantly granulocytes, and not on lymphoid or erythroid cells. In the periphery, it is expressed on granulocytes. The mAb RB6-8C5 (Cat. no. 557445/553123) recognizes both Ly-6G and Ly-6C and blocks the binding of mAb 1A8 to Ly-6G.
Spécification
Ly-6G | |
Monoclonal | |
0.2mg/mL | |
Ly6g; Lymphocyte antigen 6G; Lymphocyte antigen 6 complex, locus G; Gr1 | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
1A8 | |
Alexa Fluor 700 | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Ly-6G-transfected EL4J Cell Line | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |