Learn More
anti-GALT, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Marque:  Abnova H00002592-PW1.1Set
Code produit. 16035378
  • 809.00€ / 1 kit
Découvrez plus d'offres spéciales
Includes: Antibody pair for IP: rabbit polyclonal anti-GALT (300μL), Antibody pair for WB: mouse purified polyclonal anti-GALT (50μg)
Spécification
GALT
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
GALT
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-GALT (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-GALT (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Immunoprecipitation, Western Blot
Unconjugated
GALT
Mouse, Rabbit
RUO
2592
Human
Liquid
Promotions
For Research Use Only