Learn More
EPH receptor B2, Mouse, Clone: 1F10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant EPHB2.
Marque: Abnova H00002048-M01A.200uL
Description
Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. The protein encoded by this gene is a receptor for ephrin-B family members. [provided by RefSeq
Sequence: CIANAEEVDVPIKLYCNGDGEWLVPIGRCMCKAGFEAVENGTVCRGCPSGTFKANQGDEACTHCPINSRTTSEGATNCVCRNGYYRADLDPLDMPCTTIPSpécification
EPH receptor B2 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
NM_017449 | |
EPHB2 | |
EPHB2 (NP_059145, 226 a.a. ∼ 325 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
Yes | |
2048 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
1F10 | |
Mouse monoclonal antibody raised against a partial recombinant EPHB2. | |
EPHB2 | |
CAPB/DRT/EPHT3/ERK/Hek5/MGC87492/PCBC/Tyro5 | |
Mouse | |
200 μL | |
Kinases | |
Primary | |
Human | |
Antibody | |
IgM κ |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.