missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CDK2, CCNA1 Protein Protein Interaction Antibody Pair, Abnova™
Marque: Abnova DI0407.1Set
Includes: CDK2 rabbit purified polyclonal antibody (20μg); CCNA1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the CDK2 protein, and the other against the CCNA1 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Spécification
CDK2 | |
Monoclonal/Polyclonal | |
CDK2 | |
CDK2 | |
1 Set | |
Primary | |
Antibody pair set content: 1. CDK2 rabbit purified polyclonal antibody (20 ug) 2. CCNA1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
p33(CDK2) | |
Mouse, Rabbit | |
RUO | |
1017/8900 | |
Human | |
CDK2 & CCNA1 Protein Protein Interaction Antibody Pair |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu