missing translation for 'onlineSavingsMsg'
Learn More

CD3 Mouse anti-Human, Brilliant Violet 605, Clone: SK7, BD

Mouse Monoclonal Antibody

164.00€ - 381.00€

Spécification

Antigène CD3
Clone SK7
Applications Flow Cytometry
Classification Monoclonal
Conjugué Brilliant Violet 605
Consulter d'autres spécifications

Produits 2
Product Code Brand Quantité Price Quantity & Availability  
Product Code Brand Quantité Price Quantity & Availability  
15852609
missing translation for 'viewDocuments'
BD Biosciences
563217
25 Tests
164.00€
1 pièce
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
15872609
missing translation for 'viewDocuments'
BD Biosciences
563219
100 Tests
381.00€
1 pièce
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

The SK7 (Leu-4) monoclonal antibody specifically binds to the epsilon chain of the CD3 antigen/T-cell antigen receptor (TCR) complex. This complex is composed of at least six proteins that range in molecular weight from 20 to 30 kDa. The antigen recognized by CD3 antibodies is noncovalently associated with either α / β or γ/ δ TCR (70 to 90 kDa). The CD3 antigen is present on 61% to 85% of normal peripheral blood lymphocytes 60% to 85% of thymocytes and on Purkinje cells in cerebellum. The soluble form of this antibody has a mitogenic effect on most peripheral blood T lymphocytes, provided appropriate functional monocytes are present.

This antibody is conjugated to BD Horizon BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max of 602-nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20-nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605-nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser.

Specifications

Specifications

CD3
Flow Cytometry
Brilliant Violet 605
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
CD3-epsilon; CD3E; Leu4; T-cell surface antigen T3/Leu-4 epsilon chain; T3E
SK7
Monoclonal
Mouse
Human
Human Thymocytes
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Certificates

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CD3 Mouse anti-Human, Brilliant Violet 605, Clone: SK7, BD

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.