missing translation for 'onlineSavingsMsg'
Learn More

CD14 Mouse anti-Human, FITC, Clone: M5E2, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 555397

 Afficher plus de versions de ce produit

Code produit. 15808138

  • 371.00€ / 1 pièce

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Découvrez plus d'offres spéciales
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Description

Description

Reacts with the 53-55kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody
development only or reported in the literature.

Host Species: Mouse
Clone: M5E2
Isotype: IgG2a, κ
Cross Reactivity: Human, Rhesus, Cynomolgus, Baboon, Dog

Flow Cytometry

TRUSTED_SUSTAINABILITY
Spécification

Spécification

CD14
Monoclonal
FITC
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
M5E2
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human CD14 Protein
100 Tests
Immunology
Baboon, Canine, Cynomolgus, Human, Rhesus Monkey
IgG2a κ
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

For Research Use Only.