Learn More
CD11b/Mac-1 Mouse anti-Human, APC, Clone: ICRF44, BD
Mouse Monoclonal Antibody
262.00€ - 580.00€
Spécification
Antigène | CD11b |
---|---|
Clone | ICRF44 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugué | APC |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15830689
|
BD Biosciences
561015 |
25 Tests |
262.00€
1 pièce |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
15856168
|
BD Biosciences
550019 |
100 Tests |
580.00€
1 pièce |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Reacts with the 165kDa adhesion glycoprotein (Mac-1, integrin αM subunit) which associates with the 95kDa integrin β2 (CD18) to form the CD11b/CD18 complex. CD11b/Mac-1 is expressed on activated lymphocytes, monocytes, granulocytes, and a subset of NK cells. CD11b functions in cell-cell and cell-substrate interactions and as a receptor for iC3b, CD54 (ICAM-1), CD102 (ICAM-2) and CD50 (ICAM-3). This antibody significantly inhibits PMN aggregation in response to fMLP. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: ICRF44
Isotype: IgG1, κ
Cross Reactivity: Human
Flow Cytometry
Spécification
CD11b | |
Flow Cytometry | |
APC | |
RUO | |
Human | |
Human monocytes | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
ICRF44 | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
MAC-1A; Mac-1; ITGAM; Integrin alpha M; CR3A; CR-3 alpha; Mo1; SLEB6 |
For Research Use Only.