Learn More
Angiopoietin 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 23302-1-AP-150UL
 Afficher plus de versions de ce produit
Code produit. 16892275
  • 410.97€ / 150µL
Découvrez plus d'offres spéciales
Description
Description
Angiopoietins are proteins with important roles in vascular development and angiogenesis. All angiopoietins bind with similar affinity to an endothelial cell-specific tyrosine-protein kinase receptor. The protein encoded by this gene is a secreted glycoprotein that activates the receptor by inducing its tyrosine phosphorylation. It plays a critical role in mediating reciprocal interactions between the endothelium and surrounding matrix and mesenchyme. The protein also contributes to blood vessel maturation and stability, and may be involved in early development of the heart.
Spécification
Spécification
Angiopoietin 1
Polyclonal
Unconjugated
ANGPT1
AGP1, AGPT, ANG 1, ANG1, angiopoietin 1, ANGPT1, KIAA0003
Rabbit
Antigen Affinity Chromatography
RUO
11600, 284, 89807
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O08538, O35460, Q15389
ANGPT1
Angiopoietin 1 Fusion Protein Ag18829
150 μL
Primary
Human, Mouse, Rat