missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiogenin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA5139827
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: QDNSRYTHFL TQHYDAKPQG RDDRYCESIM RRRGLTSPCK DINTFIHGNK RSIKAICENK NGN
Angiogenin (ANG or ANG I) is important for the process of neovascularization and formation of new blood vessels. ANG is similar to pancreatic ribonuclease A and functions as a tRNA-specific ribonuclease that abolishes protein synthesis by specifically hydrolyzing cellular tRNAs. It interacts with endothelial cell-surface actin and may cause changes in the cell cytoskeleton. ANG is thought to be involved in the development of solid tumors and its antagonists are capable of inhibiting tumor growth. Defects in ANG are the cause of susceptibility to amyotrophic lateral sclerosis type 9 (ALS9). Angiogenin is a genetic link between ALS and PD.Spécification
Angiogenin | |
Polyclonal | |
Unconjugated | |
ANG | |
AI385586; ALS9; ANG; Ang1; Angiogenin; angiogenin ribonuclease 1; angiogenin, ribonuclease A family, member 1; angiogenin, ribonuclease, RNase A family, 5; angiogenin-1; epididymis luminal protein 168; HEL168; MGC22466; MGC71966; RAA1; Ribonuclease 5; ribonuclease A A1; ribonuclease A family member 5; RNase 5; RNASE4; RNASE5; Rnase5a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
283 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 μg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P03950 | |
ANG | |
Recombinant protein corresponding to Human ANG | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu