missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha-Smooth Muscle Actin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA137024
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody is predicted to react with bovine, chicken, amphibian, mouse, porcine, rabbit and rat based on sequence homology. A suggested positive control is small intestine (smooth muscle) or leiomyosarcoma.
Smooth Muscle Actin belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actin being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. In particular, smooth muscle actin is an alpha actin that is found in skeletal muscle. Actin exists as a ubiquitous protein involved with filament formation that make up large portions of the cytoskeleton. Actin filaments interact with myosin to assist in muscle contraction as well as aiding in cell motility and cytokinesis. Smooth muscle actin is found on smooth muscle vessel walls, gut wall, myometrium, myoepithelial cells in breast and salivary glands. Defects in the smooth muscle actin gene cause aortic aneurysm familial thoracic type 6. Actin isoforms differ slightly in their N-terminus and the sequences of each are perfectly conserved in higher vertebrates. Alpha-smooth muscle actin is abundant in vascular and visceral smooth muscle cells. In addition, it has also been shown that smooth muscle actin appear in stress fibers of fibroblastic cells during pathological situations involving contractile phenomena such as wound healing and fibrocontractive diseases. Multiple alternatively spliced variants of smooth muscle actin have been identified.
Spécification
Alpha-Smooth Muscle Actin | |
Polyclonal | |
ACTA2 | |
AAT6, ACTSA, alpha-sma | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
59 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
P62736 | |
ACTA2 | |
Synthetic peptide from N-terminus of human alpha smooth muscle isoform of actin | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Alpha-Smooth Muscle Actin Polyclonal Antibody, Invitrogen™ > 1 mL; Unconjugated
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu