missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Adaptin Monoclonal Antibody (AP6), Alexa Fluor™ 488, Invitrogen™
Mouse Monoclonal Antibody
Marque: Thermo Scientific MA1064A488
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Mouse Monoclonal Antibody
alpha Adaptin Monoclonal antibody specifically detects alpha Adaptin in Human samples. It is validated for Immunocytochemistry, ImmunofluorescenceSpécification
alpha Adaptin | |
Monoclonal | |
1.0 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide; pH | |
O95782 | |
Ap2a1 | |
Purified adaptors. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence | |
AP6 | |
Alexa Fluor 488 | |
Ap2a1 | |
100 kDa coated vesicle protein A; Adaptor protein complex AP-2 subunit alpha-1; Adaptor-related protein complex 2 subunit alpha-1; Adtaa; ADTAB; Alpha1-adaptin; alpha2-adaptin; Alpha-adaptin A; alpha-adaptin C; AP2; AP-2 complex subunit alpha-1; Ap2a1; AP2-ALPHA; BOS_17797; Clapa1; CLAPA2; Clathrin assembly protein complex 2 alpha-A large chain; HIP9; HIP-9; HYPJ; Plasma membrane adaptor HA2/AP2 adaptin alpha A subunit | |
Mouse | |
Protein G | |
RUO | |
160 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |