missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldosterone Antibody, Alexa Fluor™ 594, Novus Biologicals™
Sheep Polyclonal Antibody
Marque: Novus Biologicals NB100-62320AF594
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Aldosterone Polyclonal antibody specifically detects Aldosterone in All samples. It is validated for ELISASpécification
Aldosterone | |
Polyclonal | |
50mM Sodium Borate | |
Made to Broad ALDOSTERONE | |
0.1 mL | |
Lipid and Metabolism | |
All species | |
Purified |
ELISA | |
Alexa Fluor 594 | |
Sheep | |
Immunogen affinity purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG |