Learn More
Tube Al-23, une extrémité fermée, dia. ext. : 15 mm, dia. int. de : 10mm, Thermo Scientific™
98.80€ - 222.00€
Spécification
Nom chimique ou matériau | Al-23 Tube |
---|---|
Note relative au nom | One end closed |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
11392768
|
Thermo Scientific Alfa Aesar
030074.DM |
100 mm | N/A | ||||||
11352768
|
Thermo Scientific Alfa Aesar
030074.DR |
200 mm | N/A | ||||||
11362768
|
Thermo Scientific Alfa Aesar
030074.DW |
300 mm | N/A | ||||||
11382768
|
Thermo Scientific Alfa Aesar
030074.EH |
600 mm | N/A | ||||||
11372768
|
Thermo Scientific Alfa Aesar
030074.EM |
800 mm | N/A | ||||||
11302778
|
Thermo Scientific Alfa Aesar
030074.EO |
1000 mm | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécification
Al-23 Tube | |
One end closed |
Sécurité et traitement
missing translation for 'tsca' : U
Recommended Storage : Température ambiante