Learn More
Advanced Instruments Osmometer Standards
For in vitro diagonostic use
Marque:  Advanced Instruments 3MA020
Code nomenclature Nacres: LB.42
Informations supplémentaires : Poids : 0.02000kg
Code produit. 15138616
  • 110.72€ / 10 pièces
Expédition estimée: 06-08-2024
pour voir le stock disponible 
Description
Description
For in vitro diagonostic use
  • 2-Hydroxybiphenyl sodium salt tetrahydrate
  • 1% added as preservative
Spécification
Spécification
2°C to 30°C
2- Hydroxybiphenyl Sodium Salt Tetrahydrate, 0.01% added as preservative
To check the performance of your osmometer and test methods, in-vitro diagnostic use
10 x 2 mL