Learn More
Acetoacetyl CoA synthetase Antibody (011), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Marque: Novus Biologicals NBP2-89801PCP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Acetoacetyl CoA synthetase Monoclonal antibody specifically detects Acetoacetyl CoA synthetase in Human samples. It is validated for ELISA
Spécification
| Acetoacetyl CoA synthetase | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 65985 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| 011 | |
| ELISA | |
| acetoacetate-CoA ligase, acetoacetyl-CoA synthetase, ACSF1FLJ41251, Acyl-CoA synthetase family member 1, EC 6.2.1, EC 6.2.1.16, FLJ12389, homolog of C. elegans supressor of ras 5 (sur-5), Protein sur-5 homolog, SUR-5 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Acetoacetyl CoA synthetase (Accession#: NP_076417.2; Met1-Phe672). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |